Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Reaction Vessels
Reaction Vessels, Cylindrical Shape
Accessories for pressure vessels
Accessories for pressure vessels
Accessories for pressure vessels
Catalog # 516-9054
Supplier:
Cytiva (Formerly Pall Lab)
Expand image
Accessories for pressure vessels
Catalog # 516-9054
Supplier:
Cytiva (Formerly Pall Lab)
Specifications
Description:
Pressure vessel replacement parts kit, includes pressure gauge, safety relief valve and all necessary connections
Pk:
1
Specifications
About this item
Product Details
Product Details