

Iodine ≥99.99% (metals basis), crystalline
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Merck Index:13,05036
Boiling Pt:184.3 °C (1013 hPa)
Melting Pt:183…186 °C (subl.)
Density:4.930
Storage Temperature:Ambient
EINECS:231-442-4
UN:3495
ADR:8,III
:GHS07
:GHS09
Related Information
Frequently Bought Together