Order Entry
Slovenia
ContactUsLinkComponent
Calcium standard solution, 1,000 mg/L Ca in dil. nitric acid standard for ICP
Calcium standard solution, 1,000 mg/L Ca in dil. nitric acid standard for ICP
Catalog # PROOU-ICP-020
CAS Number:  
Calcium standard solution, 1,000 mg/L Ca in dil. nitric acid standard for ICP
Catalog # PROOU-ICP-020
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Environmentally Preferable:
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item