VWR® Absorbent Bench Underpads
Supplier: VWR Collection
Certificates
About this item
Absorbent underpads with a three-layer construction made from PP top and back sheets and a cellulose filler.
- Top sheet made from white spunbound polypropylene
- Absorbent cellulose fluff filler between two layers of tissue for superior wicking of fluid
- Cushion effect to protect labware from breakage
- Embossed PP backing sheet comes in different colours
- Sealed on four sides to prevent liquid seepage
Deluxe pads are more absorbent and thicker with more cellulose fluff than standard pads.
Ordering information: Super Deluxe pads have an added polymer that absorbs more moisture.
Frequently Bought Together