Penicillinase (beta-Lactamase)
Glass bottle, 1 mL
Catalog # FITZ20-PR92
Supplier: FITZGERALD INDUSTRIES INTL
Penicillinase (beta-Lactamase)
Catalog # FITZ20-PR92
Supplier: FITZGERALD INDUSTRIES INTL
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Cat. No.:FITZ20-PR92
- Storage conditions:Refrigerator
- Enzyme name:Penicillinase (beta-Lactamase)
- CAS No.:9001-74-5
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



