ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Catalog # ROCH10128031001
Supplier: ROCHE
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:787.37 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
EINECS:246-129-8
CAS Number:24292-60-2
Formula:C₂₁H₂₆N₇Na₂O₁₇P₃
MDL Number:MFCD00065390