Order Entry
Slovenia
ContactUsLinkComponent
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
  ROCH10240354001
 :  ROCHE
 :  
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
  ROCH10240354001
 :  ROCHE
 :  
  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
 :
 :787.37 g/mol
 :M-Clarity™ quality level = MQ100
 :Freezer
 :246-129-8
 :24292-60-2
 :C₂₁H₂₆N₇Na₂O₁₇P₃
 :MFCD00065390