Methyl violet 2B, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
CAS Number:8004-87-3
:
Formula:C₂₄H₂₈ClN₃
MDL Number:MFCD00012568
Molecular Weight:393.96 g/mol
Melting Pt:137
ADR:9 III
:M-Clarity™ quality level = MQ100
UN:3077
:GHS05
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together