Order Entry
Slovenia
ContactUsLinkComponent
 
Litmus, pure indicator grade
 
undefined
Litmus, pure indicator grade
 

 

  • Appearance:
    Dark blue to black Powder or granules
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.01 % in water) Opaque dark blue
  • Visual transition interval:
    from pH 4.8 (red) to pH 8.3 (blue)

 

 

Frequently Bought Together