Order Entry
Slovenia
ContactUsLinkComponent
 
Deionisers, Micromeg
  171-0484
undefined
Deionisers, Micromeg
  171-0484

 

  • Description:
    Deioniser, Micromeg
  • Environmentally Preferable:
  • W×D×H:
    200×230×550 mm
  • Max. flow rate:
    90 l/h
  • Weight:
    6,5 kg
  • Pk:
    1

 

 

This wall mounted disposable deioniser produces water up to a quality of 10M.-cm at flows of up to 90 l/hr. Designed for users with a requirement for low volumes deionised water, the Micromeg requires no electrical input as its water quality meter is battery operated.

  • Disposable cartridge
  • New cartridge can be installed in seconds