Order Entry
Slovenia
ContactUsLinkComponent
Dissecting ray with wax
Dissecting ray with wax
Catalog # 761-0036
Supplier:  BREUKHOVEN
undefined
Dissecting ray with wax
Catalog # 761-0036
Supplier:  BREUKHOVEN

Specifications

  • W×D×H:
    265×162×20 mm
  • Pk:
    1

Specifications

About this item

Stainless steel with wax.

  • Working area: 135×237 mm
  • Reusable by melting the wax filling
  • High quality

For use in laboratories and schools.