Centrifuges, benchtop, ventilated/heated/refrigerated, 6-16 / 6-16H / 6-16K
: Sigma
Universal benchtop centrifuges with speed range up to 13500 min⁻¹, and also suitable for low speeds e.g. for cell sedimentation (accuracy ±1 min⁻¹).
- Maintenance-free induction drive motor; A microcontroller controls speed, time, temperature and relative centrifugal force (RCF)
- Control unit Spincontrol Comfort
- Lid can easily be opened due pneumatic swing support
- Motorised lid lock
- Safety features: Magnetic rotor identification preventing the rotor from overspeeding, imbalance switch, no need to open the casing for emergency lid lock release, control unit lockable, code protection
6-16H: Heated version of 6-16 with:
- Max. rotor temperature: +60 °C
- Special for ASTM crud oil centrifugation max. +70 °C
6-16K: Refrigerated version of 6-16 with:
- Speed range up to 15000 min⁻¹
- Possibility of precooling the rotors during standstill
- Refrigerant CFC-free
Warranty: 1 year (centrifuges), 2 years (accessories).
Warranty: one year (centrifuges), two years (accessories)
: Rotors and accessories available separately.
Frequently Bought Together