Centrifuge, benchtop, refrigerated, high speed, 3-30 K
Supplier: Sigma
Certificates
About this item
High speed refrigerated benchtop centrifuge for centrifugal forces of up to and more than 60000×g.
- Simple operation, single-knob control with LCD-display
- Efficient cooling system, pre-selection from –20 to +40 °C
- Motorised (soft close) lid lock system
- Maintenance-free brushless drive motor, controlled by a microcontroller
- Magnetic rotor identification preventing overspeeding rotors
Warranty: 1 year (centrifuges), 2 years (accessories).
Ordering information: Rotors and accessories available separately.
Related Information
Product Information
Frequently Bought Together