Precision balances, Education Series
Supplier: Mettler - Toledo
Certificates
About this item
The Education series are simple precision balances designed for the needs of the classroom with a robust housing and clear LCD. The range of applications includes % weighing and Recall weight value. Alongside the usual units of measure kg, g and mg is the unique Newton Factor where the balance can be adjusted according to the geographical location. Below balance weighing is available on all models.
- Internal calibration adjustment at the touch of a button on EL-IC models
- Calibration by manual adjustment with external weights on standard models
- RS232 for connection to printer or PC
Models with glass draught shields with sliding doors have free access from right left and above.
Frequently Bought Together