PCR plates, 24-, 32- and 48-well, Axygen®
: Corning
PP. Designed to fit 96-well thermal cycler blocks.
- Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
- Certified free from DNase, RNase and endotoxins for highest sample integrity
These multiwell plates are specifically designed for PCR applications and are ideal for thermal transfer application. Max. RCF 4000 g.
- Catalog No:
- 732-0903
- 732-0904
- 732-0905
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Frequently Bought Together