Bottle, narrow neck, round, PP, with screw caps
Supplier: KARTELL
Certificates
About this item
In PP, with threaded cap.
- With standardised GL thread
- Perfect sealing
Certifications: Compliant with DIN 13316 and DIN 168.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together