Compressed air sprayer
215-6575
- Type:with short nozzle
- Capacity:1000 ml
- Pk:1
HDPE.
- Good chemical resistance, good temperature resistance
- With screw threaded bottle
- Choose between fine mist or spray by adjusting the spray nozzle
For aqueous solutions. Air is pumped into the container using a pump lever (depending on filling level approx. 20 to 30 pump actions). The spray mist or jet can be easily regulated with the spray nozzle.