Order Entry
Slovenia
ContactUsLinkComponent
Sulphomolybdic reagent R2 Reag. Ph. Eur. 1086400
Corrosive
Environment
Sulphomolybdic reagent R2 Reag. Ph. Eur. 1086400
Catalog # 87946.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Environment
Sulphomolybdic reagent R2 Reag. Ph. Eur. 1086400
Catalog # 87946.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Storage Temperature:Ambient
EINECS:234-722-4
UN:1830
ADR:8,II
 :GHS05
 :GHS09
CAS Number:12054-85-2
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00011596