Order Entry
Slovenia
ContactUsLinkComponent
Pararosaniline solution Reag. Ph. Eur. 1062201, decolourised
Health
Pararosaniline solution Reag. Ph. Eur. 1062201, decolourised
Catalog # 87885.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Pararosaniline solution Reag. Ph. Eur. 1062201, decolourised
Catalog # 87885.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :305.38 g/mol
Boiling Pt:~100 °C (1013 hPa)
Density:1.01 g/cm³ (20 °C)
 :GHS08
Storage Temperature:Ambient
CAS Number:467-62-9
 :25620-78-4
Formula:C₁₉H₁₉N₃O
MDL Number:MFCD00036222