
Titanium trichloride sulphuric acid reagent Reag. Ph. Eur. 1091202
Catalog # 87711.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:240.01 g/mol
Storage Temperature:Ambient
UN:2796
ADR:8,II
:GHS05