IPTG (isopropyl-β-D-thiogalactopyranoside), Sigma-Aldrich®
Catalog # ROCH10724815001
Supplier: ROCHE
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:109.5…110.5 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
EINECS:206-703-0
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273