N-Acetyltryptamine 98%
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:1016-47-3
:
Formula:C₁₂H₁₄N₂O
MDL Number:MFCD00209910
:202.2524 g/mol
Boiling Pt:484.7°C
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to light yellow to light orange crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:74°C to 78°C
- Water:≤2 %
Specification Test Results
Frequently Bought Together