Order Entry
Slovenia
ContactUsLinkComponent
 
Magnesium silicate for chromatography
Catalog # MANA815710.5
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Magnesium silicate for chromatography
Catalog # MANA815710.5
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :100.38 g/mol
EINECS:215-681-1
CAS Number:1343-88-0
Formula:MgO₃Si
MDL Number:MFCD00078249