Specifications
- W×D×H:110×130×660 mm
- Electrical:230 V, 50 Hz
- Power consumption:110 W (0,5 A)
- Pk:1
Specifications
About this item
Operation principleis based on a constant, forced air circulation through recirculator's chamber in close vicinity to UV lamps, thusensuring maximalefficiency of disinfection. Theinner mirror surface of recirculator chamber reflects ultraviolet rays thereby increasing the density of UV radiation and enhancing the disinfection effect.
- Recirculators are effective against common airborne diseases by disinfecting the air and efficiently destroying disease-causing agents (viruses, microorganisms) by UV radiation
- UVR-Mi is the more powerful model of UV air flow cleaner with two UV-lamps
- Provide complete protection from UV radiation
- Easy to install, operateand maintain
- Very low noise level
- Built-in timer allows to control the UV lamp operating time
- Digital control unit allows to track overall UV lamp operating time
UV Air Recirculators are ideal for air disinfection in hospitals (especially in outpatient departments, operating rooms, emergency rooms, delivery rooms and more), kindergartens, research laboratories, veterinary clinics. UV Air Flow Cleaner-Recirculator consists ofa germicidal UV lamps, a fan unitequipped with dust filtersand a control unit, confined in a flowthrough chamber.
Recirculator fixation:
Convenient fixation on walls (standard)
Mounting on a movablestand (optional)
UV radiation source: 2 lamps - 25 W, bactericidal UV-C, TUV25W 1SL/25
UV radiation level: 36 mW/cm²/s
Radiation type: UV (λ=253,7 nm), ozone-free
Air-flow productivity: 29 m³/h
Lamp service life: 9000 h
Display: LCD
Timer: 1 min to 24 h/non-stop
Tripod UVR-S dimensions: 510×510×~1100 mm