Order Entry
Slovenia
ContactUsLinkComponent
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99%, pure
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99%, pure
Certificates
undefined
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99%, pure
Certificates

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤1 % (K.F.)
  • HPLC:
    ≥98.5 %
  • Specific optical rotation:
    -60° to -64° (20°C, 589 nm) (c=1, H2O/DMF 1/1)

Specification Test Results

Related Information

Frequently Bought Together