
Nitro blue tetrazolium chloride (NBT), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:298-83-9
:
Formula:C₄₀H₃₀Cl₂N₁₀O₆
MDL Number:MFCD00012159
:817.65 g/mol
Melting Pt:200 °C
:M-Clarity™ quality level = MQ100
EINECS:206-067-4
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together