Human Serum AB (Off The Clot), Aseptic Assurance System
GEMB100-318-750
New Product
- Serum format:Frozen
- Sterility:Sterile filtered
- Description:Human serum AB (off the clot) - AAS
- Shipping conditions:Ships on dry ice
- Origin:US
- Storage temperature:Store frozen at −20...−10 °C
- Pk Info:200×100 ml
- Pack type:BAGA
- Pk:100 mL
Product has minimum order quantity of 200 units. Off the clot human serum AB is collected from normal healthy male donors at FDA-licensed facilities in the United States. As a natural off the clot product, this material contains no anticoagulants/preservatives. All donor units are tested for viral markers and found to be non-reactive.
- Supports clinical and commercial applications
- Aseptic assurance system