Order Entry
Slovenia
ContactUsLinkComponent
Magicard™ Monochrome Black Film + Overcoat for Magicard™ 300
Magicard™ Monochrome Black Film + Overcoat for Magicard™ 300
Catalog # BRDY321974
Supplier:  Brady
New Product
undefined
Magicard™ Monochrome Black Film + Overcoat for Magicard™ 300
Catalog # BRDY321974
Supplier:  Brady
New Product

Specifications

  • Colour:
    Transparent/black
  • Description:
    Black monochrome film with overcoat, 2-colour ribbon, 1000 single-sided cards
  • Length:
    12 m
  • Seq:
    03
  • Pk:
    1
  • Width:
    60 mm
  • For:
    Magicard™ 300

Specifications

About this item

Magicard™ 300 black monochrome ribbon with overcoat.

  • Create extra durable cards
  • Includes a cleaning roller for dust-free printing

Black mono colour dye film with overcoat protection.

Delivery: Includes 1 dye film and 1 cleaning roller.