Order Entry
Slovenia
ContactUsLinkComponent
Magicard™ Scratch-Off Film
Magicard™ Scratch-Off Film
Catalog # BRDY321964
Supplier:  Brady
New Product
undefined
Magicard™ Scratch-Off Film
Catalog # BRDY321964
Supplier:  Brady
New Product

Specifications

  • Colour:
    Silver
  • Description:
    Magicard™ scratch-off film, single colour ribbon, 1000 single-sided cards
  • Length:
    10 m
  • Pk:
    1
  • Width:
    60 mm
  • For:
    Magicard™ 300, Magicard™ 600, Pronto 100

Specifications

About this item

The Magicard™ Scratch-Off Ribbon is a grey dye film that can be scratched off the card surface.

  • High-quality, monochrome ribbon guarantees to print 1000 single-sided cards
  • Includes a cleaning roller for dust-free printing

The Magicard™ Scratch-Off Ribbon is a grey dye film that can be scratched off the card surface.

Delivery: Includes 1 dye film and 1 cleaning roller.