Order Entry
Slovenia
ContactUsLinkComponent
Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 3F9]
Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 3F9]
Supplier:  ANTIBODIES.COM
New Product
Certificates
undefined
Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 3F9]
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Catalog No:
  • ANTIA85327-50
  • ANTIA85327-100
  • Antigen symbol:
  • CRISPR-Cas9
  • CRISPR-Cas9
  • Antigen name:
  • Cas9
  • Cas9
  • Antigen synonyms:
  • CRISPR-associated endonuclease Cas9/Csn1|SpyCas9|CRISPR-Cas9/Csn1|CRISPR-Cas9
  • CRISPR-associated endonuclease Cas9/Csn1|SpyCas9|CRISPR-Cas9/Csn1|CRISPR-Cas9
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 3F9
  • 3F9
  • Reactivity:
  • S. pyogenes
  • S. pyogenes
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID# Q99ZW2
  • UniprotID# Q99ZW2
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant fragment corresponding to amino acids 1-608 of Streptococcus pyogenes Cas9 (CDJ55032.1), expressed in and purified from E. coli
  • Recombinant fragment corresponding to amino acids 1-608 of Streptococcus pyogenes Cas9 (CDJ55032.1), expressed in and purified from E. coli
  • Form:
  • Liquid
  • Liquid
  • Molecular weight:
  • 160 kDa
  • 160 kDa
  • Purification:
  • Immunogen affinity purification
  • Immunogen affinity purification
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage buffer:
  • Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
  • Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
  • Storage temperature:
  • Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C
  • Tested applications:
  • ICC/IF, IHC
  • ICC/IF, IHC

Specifications

Frequently Bought Together