Order Entry
Slovenia
ContactUsLinkComponent
Anti-Myeloperoxidase Rabbit Recombinant Antibody [Clone: MPO/33R]
Anti-Myeloperoxidase Rabbit Recombinant Antibody [Clone: MPO/33R]
Catalog # ANTIA278030-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Myeloperoxidase Rabbit Recombinant Antibody [Clone: MPO/33R]
Catalog # ANTIA278030-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    84 kDa myeloperoxidase
  • Antigen symbol:
    Myeloperoxidase
  • Clonality:
    Recombinant
  • Clone:
    MPO/33R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05164
  • Antigen synonyms:
    fj80f04|myeloid-specific peroxidase|mpx|89 kDa myeloperoxidase|Myeloperoxidase light chain|Myeloperoxidase heavy chain|MPO|EC 1.11.1.7|EC1.11.2.2
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    heavy - light promoter: 72 kDa/dimer: 140 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 150-250, of human Myeloperoxidase. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [MPO/33R] antibody to Myeloperoxidase for WB and IHC-P with samples derived from human.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Myeloperoxidase
Clonality: Recombinant
Clone: MPO/33R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human