Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-EPX Mouse Recombinant Antibody [clone: rEPO104]
  ANTIA253585-100
New Product
undefined
Anti-EPX Mouse Recombinant Antibody [clone: rEPO104]
  ANTIA253585-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Eosinophil peroxidase
  • Antigen symbol:
    EPX
  • Clonality:
    Recombinant
  • Clone:
    rEPO104
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P11678
  • Antigen synonyms:
    Eosinophil peroxidase heavy chain|EPER|EPO|PERE|Eosinophil peroxidase [Precursor]|EPP|EPX|PERE_HUMAN|EPX PEN
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    70 kDa (55 kDa - Heavy Chain/15 kDa - Light Chain)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human eosinophils from a patient with hypereosinophilic syndrome
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant mouse monoclonal [rEPO104] antibody to EPX for Flow Cytometry, IF and IHC-P with samples derived from human.

  • Validated Applications: Flow Cytometry, IF, IHC-P
  • Recommended Dilutions: Flow Cytometry: 0,5 to 1 µg/million cells, IF: 0,5 to 1 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: EPX
Clonality: Recombinant
Clone: rEPO104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human