Order Entry
Slovenia
ContactUsLinkComponent
Anti-AMPK beta 1 Rabbit Polyclonal Antibody
Anti-AMPK beta 1 Rabbit Polyclonal Antibody
Catalog # ANTIA95002-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-AMPK beta 1 Rabbit Polyclonal Antibody
Catalog # ANTIA95002-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    1300015D22Rik
  • Antigen symbol:
    AMPK beta 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9Y478
  • Antigen synonyms:
    AMPK beta 1 chain|AMPK|5'-AMP-activated protein kinase beta-1 subunit|AMP-activated, noncatalytic, beta-1|5''-AMP-activated protein kinase subunit beta-1|AAKB1_HUMAN|AMP-activated protein kinase beta subunit|AMPK beta 1|AMP-ACTIVATED PROTEIN KINASE, NONCATALYTIC, BETA-1
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    30 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human AMPK beta1 around the phosphorylation site of Ser181 (amino acids 147-196)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to AMPK beta1 (phospho Ser181) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, IHC, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000

Type: Primary
Antigen: AMPK beta 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat