Anti-AMPK alpha 1 Rabbit Polyclonal Antibody
Catalog # ANTIA309813-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:5 AMP activated protein kinase alpha 1catalytic subunit
- Antigen symbol:AMPK alpha 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q13131
- Antigen synonyms:ACACA kinase|AI194361|acetyl CoA carboxylase kinase|5' AMP activated protein kinase catalytic subunit alpha 1|5'-AMP-activated protein kinase catalytic subunit alpha-1|AAPK1_HUMAN|AI450832|AAPK1|5 AMP activated protein kinase catalytic alpha 1 chain
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% thiomersal
- Molecular weight:64 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of human AMPKa1.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to AMPK alpha 1 for WB with samples derived from mouse.
- Validated applications: WB
- Recommended dilutions: WB: 1:100 to 1:500
Type: Primary
Antigen: AMPK alpha 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse