Anti-Factor VII Mouse Monoclonal Antibody [clone: F7/3512]
ANTIA314947-100
New Product
- Antibody type:Primary
- Antigen name:Cf7
- Antigen symbol:Factor VII
- Clonality:Monoclonal
- Clone:F7/3512
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08709
- Antigen synonyms:coagulation factor VII (serum prothrombin conversion accelerator)|F7 deficiency|Factor VII light chain|Eptacog alfa|Coagulation factor VII|FA7_HUMAN|FVII|Factor VII heavy chain|Factor VII deficiency
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:200 kDa/80 kDa/73 kDa/50 kDa/43 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 366-466) of human F7 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [F7/3512] antibody to Factor VII for ELISA and IHC-P with samples derived from human.
- Validated applications: ELISA, IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Factor VII
Clonality: Monoclonal
Clone: F7/3512
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human