Anti-AMPK gamma 1 Rabbit Polyclonal Antibody
ANTIA96780-100
New Product
- Antibody type:Primary
- Antigen name:5' AMP activated protein kinase gamma 1 subunit
- Antigen symbol:AMPK gamma 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P54619
- Antigen synonyms:5''-AMP-activated protein kinase subunit gamma-1|AMPK subunit gamma-1|AMPKg|5' AMP activated protein kinase subunit gamma 1|AMPK gamma 1 chain|AAKG1_HUMAN|AMP activated protein kinase noncatalytic gamma 1 subunit|AMPK gamma 1|AMPK gamma1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:37 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human PRKAG1 (amino acids 26-75)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to PRKAG1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: AMPK gamma 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat