Anti-PCBP1 rabbit Polyclonal Antibody
Catalog # ANTIA306708-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Alpha-CP1
- Antigen symbol:PCBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q15365
- Antigen synonyms:HNRPX|HNRPE1|hnRNP E1|nucleic acid binding protein sub 2.3|heterogenous nuclear ribonucleoprotein X|heterogenous nuclear ribonucleoprotein E1|hnRNP-E1|Heterogeneous nuclear ribonucleoprotein E1|hnRNP-X
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
- Molecular weight:40 kDa
- Sequence:LRLVVPATQCGSLIGKGGCKIKEIRESTGAQVQVAGDMLPNSTERAITIAGVPQSVTECVKQICLVMLETLSQSPQGRVMTIPYQPMPASSPVICAGGQDR
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 100-200 of human hnRNP E1/PCBP1 (NP_006187.2).
- Tested applications:IHC
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
rabbit polyclonal antibody to PCBP1 for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200
Type: Primary
Antigen: PCBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat