Order Entry
Slovenia
ContactUsLinkComponent
Anti-SLC12A6 Goat Polyclonal Antibody
Anti-SLC12A6 Goat Polyclonal Antibody
  ANTIA83467-100
New Product
undefined
Anti-SLC12A6 Goat Polyclonal Antibody
  ANTIA83467-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ACCPN
  • Antigen symbol:
    SLC12A6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    KCC 3|Gaxp|KCC3 A|KCC 3B|KCC 3A|K-Cl cotransporter 3|KCC3|Furosemide sensitive KCl cotransporter 3|Electroneutral potassium-chloride cotransporter 3
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
  • Sequence:
    SRHIDVCSKTKE
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human SLC12A6 (internal region)
  • Tested applications:
    IHC
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Goat polyclonal antibody to SLC12A6 for ELISA, WB and IHC with samples derived from human.

  • Validated applications: ELISA, WB, IHC

Type: Primary
Antigen: SLC12A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human