Anti-EPX Mouse Recombinant Antibody [clone: rEPO104]
Catalog # ANTIA250405-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Eosinophil peroxidase
- Antigen symbol:EPX
- Clonality:Recombinant
- Clone:rEPO104
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P11678
- Antigen synonyms:Eosinophil peroxidase heavy chain|EPER|EPO|PERE|Eosinophil peroxidase [Precursor]|EPP|EPX|PERE_HUMAN|EPX PEN
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:70 kDa (55 kDa - Heavy Chain/15 kDa - Light Chain)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human eosinophils from a patient with hypereosinophilic syndrome
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rEPO104] antibody to EPX for Flow Cytometry, IF and IHC-P with samples derived from Human.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 0,5 to 1 µg/million cells, IF: 0,5 to 1 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: EPX
Clonality: Recombinant
Clone: rEPO104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human