Order Entry
Slovenia
ContactUsLinkComponent
Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: LLC/1738]
Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: LLC/1738]
Catalog # ANTIA252178-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: LLC/1738]
Catalog # ANTIA252178-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Bence Jones protein
  • Antigen symbol:
    Lambda Light Chain
  • Clonality:
    Monoclonal
  • Clone:
    LLC/1738
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01701, P01842
  • Antigen synonyms:
    Ig lambda chain C regions|BJP|IGLC 3|ig lambda-6 chain C region|IGLC 1|IGLC|IGLC 2|IGLC1|Constant region of lambda light chains
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    ~22,5 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Purified human lambda light chain
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [LLC/1738] antibody to Lambda Light Chain for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Lambda Light Chain
Clonality: Monoclonal
Clone: LLC/1738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human