Anti-SIGLEC1 Mouse Monoclonal Antibody [clone: SIGLEC10/7583]
Catalog # ANTIA317026-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD169
- Antigen symbol:SIGLEC1
- Clonality:Monoclonal
- Clone:SIGLEC10/7583
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q96LC7
- Antigen synonyms:FLJ00055|DKFZp667F058|FLJ32150|FLJ00411|dJ1009E24.1|FLJ00073|CD169 antigen|Sialic acid binding Ig like lectin 1|FLJ00051
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:77 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 500-697) of human SIGLEC10 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [SIGLEC10/7583] antibody to SIGLEC1 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: SIGLEC1
Clonality: Monoclonal
Clone: SIGLEC10/7583
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human