Order Entry
Slovenia
ContactUsLinkComponent
Anti-SIGLEC1 Mouse Monoclonal Antibody [clone: SIGLEC10/7582]
Anti-SIGLEC1 Mouse Monoclonal Antibody [clone: SIGLEC10/7582]
Catalog # ANTIA315641-100
Supplier:  ANTIBODIES.COM
New Product
Anti-SIGLEC1 Mouse Monoclonal Antibody [clone: SIGLEC10/7582]
Catalog # ANTIA315641-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD169
  • Antigen symbol:
    SIGLEC1
  • Clonality:
    Monoclonal
  • Clone:
    SIGLEC10/7582
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q96LC7
  • Antigen synonyms:
    FLJ00055|DKFZp667F058|FLJ32150|FLJ00411|dJ1009E24.1|FLJ00073|CD169 antigen|Sialic acid binding Ig like lectin 1|FLJ00051
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
  • Molecular weight:
    77 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 500-697) of human SIGLEC10 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [SIGLEC10/7582] antibody to SIGLEC1 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: SIGLEC1
Clonality: Monoclonal
Clone: SIGLEC10/7582
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human