Order Entry
Slovenia
ContactUsLinkComponent
Anti-AMPD3 Mouse Monoclonal Antibody [clone: AMPD3/901]
Anti-AMPD3 Mouse Monoclonal Antibody [clone: AMPD3/901]
Catalog # ANTIA248734-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-AMPD3 Mouse Monoclonal Antibody [clone: AMPD3/901]
Catalog # ANTIA248734-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adenosine monophosphate deaminase (isoform E)
  • Antigen symbol:
    AMPD3
  • Clonality:
    Monoclonal
  • Clone:
    AMPD3/901
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q01432
  • Antigen synonyms:
    AMP deaminase 3|Erythrocyte AMP deaminase|AMPD3|Erythrocyte specific AMP deaminase|AMPD3_HUMAN|Erythrocyte specific protein|AMP aminohydrolase|Adenosine monophosphate deaminase 3|AMP deaminase isoform E
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    ~90 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human AMDP3 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [AMPD3/901] antibody to AMPD3 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 2 to 4 µg/ml

Type: Primary
Antigen: AMPD3
Clonality: Monoclonal
Clone: AMPD3/901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human