Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Rat Oncomodulin/OCM ELISA Kit

Rat Oncomodulin/OCM ELISA Kit

Rat oncomodulin/OCM ELISA kit, 96 Tests
Catalog # ANTIA80349-96
Supplier:   Antibodies.com
Rat Oncomodulin/OCM ELISA Kit
Catalog # ANTIA80349-96
Supplier:  Antibodies.com
CAS Number:  
Rat Oncomodulin/OCM ELISA Kit
Rat Oncomodulin/OCM ELISA Kit

Rat oncomodulin/OCM ELISA kit, 96 Tests

CAS Number:  
Price
625,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Rat Oncomodulin/OCM ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Oncomodulin/OCM in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA Kit
  • Detection range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat Oncomodulin/OCM ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Oncomodulin/OCM in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Oncomodulin/OCM has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Oncomodulin/OCM present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Oncomodulin/OCM Antibody, which binds the captured Oncomodulin/OCM present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Oncomodulin/OCM captured in each well. The concentration of Oncomodulin/OCM can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Rat oncomodulin/OCM ELISA kit
  • Primary antibody reactivity:
    Rat
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    0,313 - 20 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Rat Oncomodulin/OCM ELISA kit exhibits high specificity and excellent specificity for the detection of rat Oncomodulin/OCM. No significant cross-reactivity or interference between Oncomodulin/OCM and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    ANTIA80349-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,188 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Oncomodulin/OCM
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.