Order Entry
Slovenia
ContactUsLinkComponent
Mouse Beta 2 Defensin/BD-2 ELISA Kit
Mouse Beta 2 Defensin/BD-2 ELISA Kit
Catalog # ANTIA311792-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Mouse Beta 2 Defensin/BD-2 ELISA Kit
Catalog # ANTIA311792-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    beta 2 Defensin/BD-2
  • Description:
    Mouse beta 2 Defensin/BD-2 ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    40 - 640 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    2,52 ng/L
  • Regulatory status:
    RUO

Specifications

About this item

Mouse beta 2 Defensin/BD-2 ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse beta 2 Defensin/BD-2 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse beta 2 Defensin/BD-2 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse beta 2 Defensin/BD-2 in serum, plasma or other biological fluids. An antibody specific for beta 2 Defensin/BD-2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the beta 2 Defensin/BD-2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-beta 2 Defensin/BD-2 antibody, which also binds the beta 2 Defensin/BD-2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-beta 2 Defensin/BD-2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-beta 2 Defensin/BD-2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of beta 2 Defensin/BD-2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of beta 2 Defensin/BD-2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.