Order Entry
Slovenia
ContactUsLinkComponent
Rat MIP-1 alpha/CCL3 ELISA Kit
Rat MIP-1 alpha/CCL3 ELISA Kit
Catalog # ANTIA74224-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Rat MIP-1 alpha/CCL3 ELISA Kit
Catalog # ANTIA74224-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    MIP-1 alpha/CCL3
  • Description:
    Rat MIP-1 alpha/CCL3 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Rat MIP-1 alpha/CCL3 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat MIP-1 alpha/CCL3. No significant cross-reactivity or interference between MIP-1 alpha/CCL3 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    31,25 - 2000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    18,75 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rat MIP-1 alpha/CCL3 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat MIP-1 alpha/CCL3 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection Range: 31,25 to 2000 pg/ml
  • Sensitivity: 18,75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat MIP-1 alpha / CCL3 ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat MIP-1 alpha / CCL3 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for MIP-1 alpha / CCL3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the MIP-1 alpha / CCL3 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-MIP-1 alpha / CCL3 Antibody, which binds the captured MIP-1 alpha / CCL3 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of MIP-1 alpha / CCL3 captured in each well. The concentration of MIP-1 alpha / CCL3 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.