Order Entry
Slovenia
ContactUsLinkComponent
Guinea Pig eNOS ELISA Kit
Guinea Pig eNOS ELISA Kit
Catalog # ANTIA73790-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Guinea Pig eNOS ELISA Kit
Catalog # ANTIA73790-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Guinea pig
  • Target protein:
    eNOS
  • Description:
    Guinea pig eNOS ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Guinea Pig eNOS ELISA kit exhibits high specificity and excellent specificity for the detection of guinea pig eNOS. No significant cross-reactivity or interference between eNOS and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    31,25 - 2000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    18,75 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Guinea Pig eNOS ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of guinea pig eNOS in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection Range: 31,25 to 2000 pg/ml
  • Sensitivity: 18,75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Guinea Pig eNOS ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of guinea pig eNOS in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for eNOS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the eNOS present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-eNOS Antibody, which binds the captured eNOS present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of eNOS captured in each well. The concentration of eNOS can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.