Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Mouse EPX ELISA Kit

Mouse EPX ELISA kit, 96 Tests
  ANTIA313860-96
Mouse EPX ELISA Kit
  ANTIA313860-96
 :  
Mouse EPX ELISA Kit
Mouse EPX ELISA Kit

Mouse EPX ELISA kit, 96 Tests

 :  
 
565,00 €
   
1 * 96 Tests
 

Mouse EPX ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse EPX in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 0,75 to 12 ng/ml
  • Sensitivity: 0,027 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse EPX ELISA kit (A313860) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse EPX in serum, plasma or other biological fluids. An antibody specific for EPX has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the EPX present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-EPX antibody, which also binds the EPX present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-EPX antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-EPX antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of EPX captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of EPX in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

 

 

  • Pk:
    96 Tests
  • Description:
    Mouse EPX ELISA kit
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma or other biological fluids
  • Sample volume:
    40 μl
  • Detection range:
    0,75 - 12 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ANTIA313860-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,027 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    EPX
  • Regulatory status:
    RUO

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.