Order Entry
Slovenia
ContactUsLinkComponent
Rat IKB alpha ELISA Kit
Rat IKB alpha ELISA Kit
Catalog # ANTIA79470-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Rat IKB alpha ELISA Kit
Catalog # ANTIA79470-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    IKB alpha
  • Description:
    Rat IKB alpha ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Rat IKB alpha ELISA kit exhibits high specificity and excellent specificity for the detection of rat IKB alpha. No significant cross-reactivity or interference between IKB alpha and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,313 - 20 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0,188 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rat IKB alpha ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat IKB alpha in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0,313 - 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat IKB alpha ELISA kit (A79470) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat IKB alpha in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for IKB alpha has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IKB alpha present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-IKB alpha Antibody, which binds the captured IKB alpha present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of IKB alpha captured in each well. The concentration of IKB alpha can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.