Order Entry
Slovenia
ContactUsLinkComponent
Rat Dehydroepiandrosterone Sulfate ELISA Kit
Rat Dehydroepiandrosterone Sulfate ELISA Kit
Catalog # ANTIA76439-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Rat Dehydroepiandrosterone Sulfate ELISA Kit
Catalog # ANTIA76439-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Dehydroepiandrosterone Sulfate
  • Description:
    Rat dehydroepiandrosterone sulfate ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Rat Dehydroepiandrosterone Sulfate ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Dehydroepiandrosterone Sulfate. No significant cross-reactivity or interference between Dehydroepiandrosterone Sulfate and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    15,625 - 1000 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    9,375 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rat Dehydroepiandrosterone Sulfate ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat Dehydroepiandrosterone Sulfate in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 15,625 to 1000 ng/ml
  • Sensitivity: 9,375 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Rat Dehydroepiandrosterone Sulfate ELISA Kit (A76439) employs the competitive enzyme immunoassay technique for the quantitative measurement of rat Dehydroepiandrosterone Sulfate in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Dehydroepiandrosterone Sulfate antigen. During the incubation, Dehydroepiandrosterone Sulfate present in the samples or standards competes with the fixed amount of immobilised Dehydroepiandrosterone Sulfate for binding sites on the Biotinylated Anti-Dehydroepiandrosterone Sulfate Antibody. The more Dehydroepiandrosterone Sulfate present in a sample or standard, the less Biotinylated Anti-Dehydroepiandrosterone Sulfate Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Dehydroepiandrosterone Sulfate Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Dehydroepiandrosterone Sulfate present in each sample or standard. The concentration of Dehydroepiandrosterone Sulfate can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.